Order Entry
Switzerland
ContactUsLinkComponent
Heptane (mixture of isomers)
Heptane (mixture of isomers)
Catalog # ROTH7725.1
Supplier:  Roth Carl
CAS Number:  
undefined
Heptane (mixture of isomers)
Catalog # ROTH7725.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
Merck Index:13,04679
Boiling Pt:97…98 °C (1013 hPa)
Melting Pt:–90.5 °C
Density:0.68 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
 :100.2 g/mol
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544