

1,3-Dichloro-2-butene cis- and trans-mixture 98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:926-57-8
:
Formula:C₄H₆Cl₂
MDL Number:MFCD00000861
:125 g/mol
Boiling Pt:125…129 °C
Melting Pt:–98 °C
Density:1.161
Flash Pt:33 °C (91 °F)
UN:1992
ADR:3,III
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
EINECS:213-138-3
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together