Glycerine solution 86-89% (by titrimetric analysis), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:56-81-5
:
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722
:92.09 g/mol
Merck Index:13,04497
Boiling Pt:290 °C (1.013 hPa)
Melting Pt:20 °C
Density:1.26 g/cm³
Flash Pt:160 °C
Storage Temperature:Ambient
EINECS:200-289-5
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating)
Specifications
Specification Test Results
- Assay:85.5 - 89.5 %
- Refractive index (n 20/D):1.453 - 1.457
- Sulfated ash:Max. 0.01 %
- Water (Karl Fischer):11 - 14 %
- Free acid (as CH₃COOH):Max. 0.002 %
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 1 ppm
- Cobalt (Co):Max. 1 ppm
- Chromium (Cr):Max. 1 ppm
- Copper (Cu):Max. 1 ppm
- Iron (Fe):Max. 1 ppm
- Potassium (K):Max. 20 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 1 ppm
- Sodium (Na):Max. 20 ppm
- Nickel (Ni):Max. 1 ppm
- Lead (Pb):Max. 1 ppm
- Zinc (Zn):Max. 1 ppm
- Ammonium (NH₄):Max. 5 ppm
- Chloride (Cl):Max. 2 ppm
- Sulfate (SO₄):Max. 5 ppm
- Reaction against H₂SO₄:complying
- Aldehydes (as HCHO):Max. 0.001 %
- Fatty acid ester:Max. 0.05 %
Specification Test Results
Frequently Bought Together