
Osmium ≥99.8% (metals basis), powder -200 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-04-2
:
Formula:Os
MDL Number:MFCD00011147
:190.23 g/mol
Storage Temperature:Ambient
EINECS:231-114-0
UN:3089
ADR:4.1,II
:GHS02
Related Information
Frequently Bought Together