Petri dishes, disposable
A cardboard disc (1,4 mm thickness) is wetted with appropriate nutrient medium before the membrane filter is placed on. They are pre-sterilised in plastic magazines, suitable for the corresponding dispenser.
- Available in two Ø
Frequently Bought Together