A range of four-place balances that incorporate a touch screen, advanced weighing and proximity sensor technologies. The easy to use control unit can be mounted on a base, above a base on an optional column, on a wall, or remotely (up to 3 metres distance with an optional cable). The touch screen display, with universal icons and informative prompts, simplifies set-up functions and basic operation. The operator has the added choice to use proximity sensors for hands-free operation when handling sensitive samples. These balances have multiple application modes from basic weighing to SQC applications, together with short stabilisation times and optimised performance specifications.
- Colour touch screen display with viewing angle adjustment in a demountable control unit
- Advanced weighing technology with stabilisation times of less than 3 seconds
- SmarText™ software with text prompts for ease of menu navigation including 'Level assist' feature with illuminated level indicator
- AutoCal™ internal calibration
- Dual-multifunctional proximity sensors for hands-free operation of basic or user-selectable functions
- LIMS compatibility
The balances feature a frameless glass draft shield with easy access, central light and removable doors for cleaning.