Order Entry
Switzerland
ContactUsLinkComponent
Copper, wire, GPR RECTAPUR®, reduced from oxide
Copper, wire, GPR RECTAPUR®, reduced from oxide
Catalog # 23028.182
Supplier:  VWR Chemicals
CAS Number:  
undefined
Copper, wire, GPR RECTAPUR®, reduced from oxide
Catalog # 23028.182
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :63.55 g/mol
Merck Index:13,02545
Boiling Pt:2595 °C (1013 hPa)
Melting Pt:1083 °C
Density:8.94 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-159-6
CAS Number:7440-50-8
Formula:Cu
MDL Number:MFCD00010965