

Copper ≥99% (metals basis), powder -625 mesh (APS 2-3,50 µm)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-50-8
:
Formula:Cu
MDL Number:MFCD00010965
:63.55 g/mol
Merck Index:13,02545
Boiling Pt:2595 °C (1013 hPa)
Melting Pt:1083 °C
Density:8.94 g/cm³ (20 °C)
UN:3089
ADR:4.1,III
:Packaged under argon.
EINECS:231-159-6
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together