Bromine, Suprapur® for trace analysis, Supelco®
1.01947.0250
: Merck
:
- Pack type:Glass bottle, plastic-coated, returnable
- Pk:250 mL
- Seq:2590
:
:159.81 g/mol
:13,01378
:58.7 °C (1013 hPa)
:–7.2 °C
:3.12 g/cm³ (20 °C)
:Ambient
:231-778-1
:1744
:8 (6.1),I
:M-Clarity™ quality level = MQ100
:GHS06
:GHS05
:GHS09
:7726-95-6
:Br₂
:MFCD00010896