Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Selenium 99.5+%, powder 200 mesh
 
undefined
 
 
Selenium 99.5+%, powder 200 mesh
 

 

  • Appearance:
    Dark grey to black Powder
  • Assay:
    ≥99.50 %
  • Particle size:
    ≤200 mesh
  • Arsenic (As):
    ≤100 ppm
  • Copper (Cu):
    ≤100 ppm
  • Iron (Fe):
    ≤200 ppm
  • Lead (Pb):
    ≤200 ppm
  • Mercury (Hg):
    ≤100 ppm
  • Nickel (Ni):
    ≤100 ppm
  • Tellurium (Te):
    ≤1000 ppm

 

 

Frequently Bought Together