Antimony ≥99.9999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
:7440-36-0
:
:Sb
:MFCD00134030
:121.76 g/mol
:12,00733
:1637 °C (1013 hPa)
:630 °C
:6.684 g/cm³ (20 °C)
:Ambient
:231-146-5
:2871
:6.1,III
:GHS07
:GHS09
Frequently Bought Together