

Antimony ≥99.5% (metals basis), powder -200 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-36-0
:
Formula:Sb
MDL Number:MFCD00134030
:121.76 g/mol
Storage Temperature:Ambient
EINECS:231-146-5
UN:2871
ADR:6.1,III
:GHS06
:GHS09
Related Information
Frequently Bought Together