


Lead ≥99.8% (metals basis), foil, Thickness: 1.19 mm (0.0469 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-92-1
:
Formula:Pb
MDL Number:MFCD00134050
:207.2 g/mol
Merck Index:12,05410
Boiling Pt:1740 °C (1013 hPa)
Melting Pt:327 °C
Density:11.3437 g/cm³ (20 °C)
EINECS:231-100-4
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together