Lead ≥99.998% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
:7439-92-1
:
:Pb
:MFCD00134050
:207.2 g/mol
:12,05410
:1740 °C (1013 hPa)
:327 °C
:11.3437 g/cm³ (20 °C)
:231-100-4
:GHS08
:GHS07
:GHS09
:Ambient
Frequently Bought Together