Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Lead 99+%, granules

Health
Irritant
Environment
Lead 99+%, granules
Certificates
Lead 99+%, granules

2 Options Available Below

Product Details & Documents
  • CAS Number:7439-92-1
  • Formula:Pb
  • MDL Number:MFCD00134050
  • Molecular Weight:207.2 g/mol
  • Merck Index:12,05410
  • Boiling Pt:1740 °C (1013 hPa)
  • Melting Pt:327.4 °C
  • Density:11,3437 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-100-4

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Grey Granules
  • Assay:
    ≥99.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 37.00
    SFr. 37.00
    1 * 100 g
       SFr. 37.00

  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 90.70
    SFr. 90.70
    1 * 500 g
       SFr. 90.70