Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Lead 99+%, powder

Lead 99+%, powder

Glass bottle for solids, 1 kg
Catalog # ACRO198110010
Health
Environment
Lead 99+%, powder
Catalog # ACRO198110010
CAS Number:  7439-92-1
Lead 99+%, powder
Lead 99+%, powder

Glass bottle for solids, 1 kg

CAS Number:  7439-92-1
Price
SFr. 83.70
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:7439-92-1
  • Formula:Pb
  • MDL Number:MFCD00134050
  • Molecular Weight:207.2 g/mol
  • Boiling Pt:1740 °C (760 mmHg)
  • Melting Pt:327.4 °C
  • Density:11,3 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-100-4
  • UN:3077
  • ADR:9,III

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Appearance:
    Grey Powder
  • Assay:
    ≥99.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.