Bottles, wide neck, with screw cap
Nome del fornitore: Brand
Certificati
Informazioni su questo articolo
PFA, translucent, with PFA screw cap.
- Maximum chemical resistance
- Heat resistant from –200 to +260 °C
- Easy to clean
High-quality bottles, specially designed for long-term storage of solvents and standards (trace analysis). PFA screw cap with integrated seal lip and buttress thread.