Funnels with short stem, ribbed
Supplier: Brand
Certificates
About this item
Borosilicate glass, ribbed walls.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For rapid filtration. Cone angle 60°. Funnel lip is ground and fire-polished.
Frequently Bought Together