ContactUsLinkComponent
Chamber slides, Corning® BioCoat™ Poly-D-lysine/laminin
Chamber slides, Corning® BioCoat™ Poly-D-lysine/laminin
Catalog # 734-1072
Supplier:  Corning
   Low Manufacturing Impact
undefined
Chamber slides, Corning® BioCoat™ Poly-D-lysine/laminin
Catalog # 734-1072
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • Description:
    Glass chamber slides
  • Material:
    PS/glass
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Pk:
    12
Specifications
About this item
These culture slides have a uniform application of poly-D-lysine/laminin.
  • Tested to promote neurite outgrowth
  • NG108 rat glioma/mouse neuroblastoma cells
  • PS vessel lid together with removal tool