ContactUsLinkComponent
Size exclusion chromatography columns, Asahipak GF-HQ series
Size exclusion chromatography columns, Asahipak GF-HQ series
Supplier:  Resonac
Certificates
Size exclusion chromatography columns, Asahipak GF-HQ series
Supplier:  Resonac
Specifications
  • Catalog No:
  • 554-1763
  • 554-1756
  • 554-1758
  • 554-1757
  • Description:
  • GF-1G 7B
  • GF-510 HQ
  • GF-7M HQ
  • GF-310 HQ
  • Max. pressure:
  • 6,5 MPa (65 bar)
  • 4,5 MPa (45 bar)
  • 7,0 MPa (70 bar)
  • pH range:
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • Temperature limit:
  • 4 - 60 °C
  • 4 - 60 °C
  • 4 - 60 °C
  • 4 - 60 °C