Laminin culture dishes, Corning® BioCoat™
Supplier: Corning
Certificates
About this item
PS coated with a uniform application of mouse laminin. Promotes cell adhesions, proliferation and differentiation of a variety of cell lines, particularly neurons, epithelial cell, myocytes and myoblasts.
Manufactured in a highly controlled environment and rigorously tested to assure product consistency and performance.
Specifications
- Catalog No:
- 734-0113
- 734-0138
- 734-0143
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 - Bedford
- ISO 14001 - Bedford