VWR® Maximum SF Boot Covers, Non Skid
Anbieter: VWR Collection
Zertifikate
Über diesen Artikel
VWR® Maximum SF boot covers are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
- Non skid
- Impervious material
- Superior friction coefficient to ensure maximum traction
- Maximum skid resistance on wet and smooth surfaces
- Clean, low linting fabric
Tested and manufactured in an ISO certified facility under stringent process controls. Lot controlled and validated through independent laboratory testing.
Length: 286 mm (M), 356 mm (U*), 381 mm (XL)
* One size fits all.
Technische Daten
- Catalog No:
- 113-9107
- 113-1187
- 113-1188
- 113-1189
- 113-1190
- 113-1191
- 113-1192
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
Technische Daten
Frequently Bought Together