VWR® Maximum SF Cleanroom Overalls
Supplier: VWR Collection
Certificates
About this item
VWR® Maximum SF overalls are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
- Available as a standard overall with collar, either elasticated ankles or non slip boot covers, or hooded overall with fluid-impervious boot covers
- Inset sleeves, zip closure, elasticated cuffs
- Superior fluid and particulate barrier
- Lowest level of particle shedding: Level 1 Helmke drum classification
- Light chemical splash resistance
- Improved breathability to ensure user comfort
- Soft, cloth-like fabric with maximum tear strength
Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
Specifications
- Catalog No:
- 113-1085
- 113-1086
- 113-1078
- 113-9161
- 113-1087
- 113-1077
- 113-1088
- 113-1089
- 113-1074
- 113-1075
- 113-1080
- 113-1076
- 113-1081
- 113-1082
- 113-1079
- 113-1084
- Model:
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Bacterial filtration efficiency:
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%