Potassium chloride saturated with silver chloride aqueous solution 3 mol/l (3 N) in aqueous solution for filling electrodes
Catalog # SCOI285138419
Supplier: SI Analytics
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
- Environmentally Preferable:
Specifications
About this item
:
:74.55 g/mol
Density:1.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl+AgCl
MDL Number:MFCD00011360