
Xylene cyanole FF ≥70% (dye content)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2650-17-1
:
Formula:C₂₅H₂₇N₂NaO₆S₂
MDL Number:MFCD00040657
:538.62 g/mol
Melting Pt:∼ 295 °C (dec.)
EINECS:220-167-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together