Order Entry
Switzerland
ContactUsLinkComponent
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), Sigma-Aldrich®
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), Sigma-Aldrich®
Catalog # 1.24644.0005
Supplier:  Merck
CAS Number:  
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), Sigma-Aldrich®
Catalog # 1.24644.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Nicotinamide adenine dinucleotide reduced, disodium salt (reduced form), NADH disodium salt, ((2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy-((((2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy)hydroxyphosphoryl)oxyphosphinic acid disodium salt
 :
 :709.41 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:210-123-3
CAS Number:606-68-8
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂
MDL Number:MFCD00036200