β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form)
Supplier: Roth Carl
Certificates
About this item
Synonyms: Nicotinamide adenine dinucleotide reduced, disodium salt (reduced form), NADH disodium salt, ((2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy-((((2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy)hydroxyphosphoryl)oxyphosphinic acid disodium salt
CAS Number:606-68-8
:
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂
MDL Number:MFCD00036200
:709.41 g/mol
EINECS:210-123-3
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together