Order Entry
Switzerland
ContactUsLinkComponent
 
Flammable
Irritant
Health
Papanicolaou's Orange G solution (OG 6)
Catalog # CTEH64072
Supplier:  CLIN TECH
CAS Number:  
Flammable
Irritant
Health
Papanicolaou's Orange G solution (OG 6)
Catalog # CTEH64072
Supplier:  CLIN TECH
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
Density:0.83 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1170
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
MDL Number:MFCD00147833