Orders LinkContactUsLinkComponent
Bouin's fluids
Bouin's fluids
Bouin's fluid, Glass bottle, 1 L
Catalog # CTEH64096/1
Marketsource
Supplier:   CLIN TECH
Bouin's fluids
Catalog # CTEH64096/1
Supplier:  CLIN TECH
Marketsource
CAS Number:  
Bouin's fluids
Bouin's fluid, Glass bottle, 1 L
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
This staining fixative is stable under normal temperature conditions and recommended use.
Bouin's fluids are used for histology and cytology applications.
Caution: May be harmful if inhaled, swallowed or absorbed through skin. May cause respiratory tract irritation.
Specifications
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    Bouin's fluid