Order Entry
Switzerland
ContactUsLinkComponent
(-)-Quinine hydrochloride dihydrate 99%
Irritant
(-)-Quinine hydrochloride dihydrate 99%
Certificates
undefined
Irritant
(-)-Quinine hydrochloride dihydrate 99%
Certificates

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or needles
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    6 % to 10 % (1 g, 105°C)
  • Sulfated ash:
    ≤0.1 %
  • Specific optical rotation:
    -245° to -258° (20°C, 589 nm) (c=2, 0.1 M HCl) (on dried substance)
  • Titration with HClO4:
    ≥98.5 %
  • Sulfate (SO4):
    ≤500 ppm

Specification Test Results

Related Information

Frequently Bought Together