
Schiff’s fuchsin-sulphite reagent, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
:
Formula:C₂₀H₂₁N₃O₃S
:383.46 g/mol
Density:1.1 g/cm³ (20 °C)
ADR:8,III
:M-Clarity™ quality level = MQ100
UN:1789
:GHS05
Specification Test Results
- Appearance (Color):Colorless
- Appearance (Form):Liquid
- Assay (Titration with Iodine):3-5 mM Sulfur dioxide per 100ml solution
- Suitable for the Detection of Glycoproteins in Polyacrylamide Gels:Suitable
Specification Test Results
Frequently Bought Together