Water deionisation cartridges, Aquadem® SDF
Supplier: ENVIROFALK PHARMAWATERSYSTEMS
Certificates
About this item
Pressure-resistant water deionisation cartridges made from stainless steel, filled with regenerable mixed-bed resins.
- Economical water deionisation for all requirements
- Achieved water quality covers the majority of common laboratory applications
- Simple handling with quick connection system
- Suitable for benchtop set up in laboratories
- Long life time, transportable
Aquadem® SDF 20: Standard equipment for laboratory glass washers.
Ordering information: Connection sets have to be ordered separately (see Accessories).
Frequently Bought Together