Order Entry
Switzerland
ContactUsLinkComponent
 
VWR® VP 820, Diaphragm Vacuum Pump
 
undefined
VWR® VP 820, Diaphragm Vacuum Pump

 

  • Catalog No:
  • 181-0072
  • Model:
  • VP 820 with CH-plug
  • Flow rate:
  • 20 l/min
  • Working overpressure (bar):
  • 1,0
  • Final pressure (abs.):
  • 8 mbar
  • Protection class:
  • IP 44
  • W×D×H:
  • 110×304×204 mm
  • Weight:
  • 9,3 kg
  • Final vacuum (abs.):
  • 8 mbar

 

Frequently Bought Together