Cold traps with/without condensation drain
For direct filling with coolant. Double-walled design ensures low coolant consumption. Includes NS 29/32 standard ground socket for connecting to the apparatus and NS 29/32 standard ground cone for connecting to the vacuum pump. DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Frequently Bought Together