Orders LinkContactUsLinkComponent
Stirrer with paddles
Stirrer with paddles
Stirrer with paddles
Stirrer with paddles
DURAN®, borosilicate glass shaft, PFTE paddles with ground and polished shaft.
 : * stirring Ø when blades extended
Product Family Options
  • NS
    Stirrer Ø
    Shaft Ø
    Shaft length
  • 29/32
    50* mm
    10 mm
    310 mm
    1 * 1 ST
    1 * 1 ST - SFr. 142.00
    SFr. 142.00
    1 * 1 ST
       SFr. 142.00
  • 29/32
    90* mm
    10 mm
    310 mm
    1 * 1 ST
    1 * 1 ST - SFr. 140.00
    SFr. 140.00
    1 * 1 ST
       SFr. 140.00
  • 29/32
    90* mm
    10 mm
    400 mm
    1 * 1 ST
    1 * 1 ST - SFr. 140.00
    SFr. 140.00
    1 * 1 ST
       SFr. 140.00
  • 29/32
    90* mm
    10 mm
    460 mm
    1 * 1 ST
    1 * 1 ST - SFr. 142.00
    SFr. 142.00
    1 * 1 ST
       SFr. 142.00
  • 29/32
    90* mm
    10 mm
    520 mm
    1 * 1 ST
    1 * 1 ST - SFr. 142.00
    SFr. 142.00
    1 * 1 ST
       SFr. 142.00