Orders LinkContactUsLinkComponent
Mesh | Avantor
Mesh
Supplier:   NEOLAB
Mesh
Supplier:  NEOLAB
Certificates
Mesh
67 Options Available Below
Product Details & Documents
Polyamide mesh (Nylon): Resistance to acids is limited and dependent on the concentration and temperature. Excellent resistance to alkalis, including caustic soda and caustic potash. Chemical performance against organic solvents is very good. Temperature resistant from 0 to 130 °C when dry, 95 to 100 °C in water.
Polyester mesh: Resistant to most acids. Not resistant to alkalis. Temperature resistant from –75 to +150 °C (for short periods), up to 125 °C for longer requirements.
PP mesh: Humidity resolution at 95% rel. air moisture and 25 °C is less than 0,01%. Resistant to alkalis, even in high concentrations. Use with organic solvents is limited. Temperature resistant from 0 to +150 °C (for short periods), up to 125 °C for longer requirements.
ETFE mesh: Humidity absorption at 20 °C and 65% relative humidity is 0%. Good resistance to acids, alkalis, solvents and hydrolysis. Temperature resistant from –190 to +150 °C when dry.
Product Family Options
Polyamide
Product Information
  • Pore size
    Availability
    Price
  • 15,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9539
  • 20,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9538
  • 25,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-2073
  • 30,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9537
  • 40,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - SFr. 77.50
    SFr. 77.50
    1 * 1 m
       SFr. 77.50
    Specifications:
    More
    Specifications
    Cat. No.
    510-9536
  • 50,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - SFr. 518.00
    SFr. 518.00
    1 * 1 m
       SFr. 518.00
    Specifications:
    More
    Specifications
    Cat. No.
    510-9535
  • 60,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - SFr. 66.20
    SFr. 66.20
    1 * 1 m
       SFr. 66.20
    Specifications:
    More
    Specifications
    Cat. No.
    510-9534
  • 70,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - SFr. 410.00
    SFr. 410.00
    1 * 1 m
       SFr. 410.00
    Specifications:
    More
    Specifications
    Cat. No.
    510-9533
  • 80,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - SFr. 62.20
    SFr. 62.20
    1 * 1 m
       SFr. 62.20
    Specifications:
    More
    Specifications
    Cat. No.
    510-9532
  • 90,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9531
  • 125,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - SFr. 52.40
    SFr. 52.40
    1 * 1 m
       SFr. 52.40
    Specifications:
    More
    Specifications
    Cat. No.
    510-9529
  • 150,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9528
  • 200,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - SFr. 410.00
    SFr. 410.00
    1 * 1 m
       SFr. 410.00
    Specifications:
    More
    Specifications
    Cat. No.
    510-9527
  • 250,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - SFr. 50.70
    SFr. 50.70
    1 * 1 m
       SFr. 50.70
    Specifications:
    More
    Specifications
    Cat. No.
    510-9526
  • 300,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9525
  • 400,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9524
  • 500,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9523
  • 600,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9522
  • 700,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9521
  • 900,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - SFr. 232.00
    SFr. 232.00
    1 * 1 m
       SFr. 232.00
    Specifications:
    More
    Specifications
    Cat. No.
    510-9485
  • 1000,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - SFr. 46.50
    SFr. 46.50
    1 * 1 m
       SFr. 46.50
    Specifications:
    More
    Specifications
    Cat. No.
    510-9484
  • 1600,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 84.80
    SFr. 84.80
    1 * 1 ROU
       SFr. 84.80
    Specifications:
    More
    Specifications
    Cat. No.
    510-9483
  • 2000,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9482
  • 3000,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9481
Polyester
Product Information
  • Pore size
    Availability
    Price
  • 10,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9517
  • 20,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9516
  • 30,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9515
  • 40,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9514
  • 53,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9513
  • 70,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9512
  • 80,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9511
  • 90,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9510
  • 105,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9509
  • 150,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9508
  • 200,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9507
  • 250,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9506
  • 300,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9505
  • 400,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9504
  • 500,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9503
  • 600,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9502
  • 700,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9501
  • 800,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9474
  • 900,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9473
  • 1000,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9472
  • 1600,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9471
  • 2000,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9470
PP
Product Information
  • Pore size
    Availability
    Price
  • 149,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU
    Specifications:
    More
    Specifications
    Cat. No.
    510-9557
  • 210,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9556
  • 295,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9554
  • 350,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9553
  • 500,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU
    Specifications:
    More
    Specifications
    Cat. No.
    510-9552
  • 710,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9551
  • 1000,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9495
  • 1200,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9494
  • 2000,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9492
  • 2400,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
    Specifications:
    More
    Specifications
    Cat. No.
    510-9491
  • 3360,00 µm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - SFr. 91.90
    SFr. 91.90
    1 * 1 m
       SFr. 91.90
    Specifications:
    More
    Specifications
    Cat. No.
    510-9490
ETFE
Product Information
  • Pore size
    Availability
    Price
  • 70,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU
    Specifications:
    More
    Specifications
    Cat. No.
    510-9561
  • 85,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU
    Specifications:
    More
    Specifications
    Cat. No.
    510-9562
  • 150,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU
    Specifications:
    More
    Specifications
    Cat. No.
    510-9568
  • 210,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    510-9569
  • 300,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU
    Specifications:
    More
    Specifications
    Cat. No.
    510-9574
  • 420,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU
    Specifications:
    More
    Specifications
    Cat. No.
    510-9575
  • 500,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU
    Specifications:
    More
    Specifications
    Cat. No.
    510-9576
  • 590,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU
    Specifications:
    More
    Specifications
    Cat. No.
    510-9577
  • 850,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU
    Specifications:
    More
    Specifications
    Cat. No.
    510-9578
  • 1000,00 µm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU
    Specifications:
    More
    Specifications
    Cat. No.
    510-9579
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..