Order Entry
Switzerland
ContactUsLinkComponent
Digital pocket refractometers, PAL series
Digital pocket refractometers, PAL series
Supplier:  Atago
Certificates
undefined
Digital pocket refractometers, PAL series
Supplier:  Atago

Specifications

  • Catalog No:
  • 635-0726
  • 635-0812
  • 635-0153
  • 635-0214
  • ATAG3810A
  • 635-0155
  • 635-0154
  • 635-0156
  • 635-0363
  • 635-0158
  • 635-0160
  • Model:
  • PAL series
  • PAL series
  • PAL series
  • PAL series
  • PAL series
  • PAL series
  • PAL series
  • PAL series
  • PAL series
  • Sample size:
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • Response time:
  • 3 s
  • 3 s
  • 3 s
  • 3 s
  • 3 s
  • 3 s
  • 3 s
  • 3 s
  • 3 s
  • 3 s
  • 3 s
  • Electrical:
  • 2 AAA batteries
  • 2 AAA batteries
  • 2 AAA batteries
  • 2 AAA batteries
  • 2 AAA batteries
  • 2 AAA batteries
  • 2 AAA batteries
  • 2 AAA batteries
  • 2 AAA batteries
  • 2 AAA batteries
  • 2 AAA batteries
  • W×D×H:
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • Weight:
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g

Specifications

Frequently Bought Together