Order Entry
Switzerland
ContactUsLinkComponent
 
Pump, suction siphon
Catalog # 181-0305
Supplier:  NRG DEVELOPMENTS
undefined
Pump, suction siphon
Catalog # 181-0305
Supplier:  NRG DEVELOPMENTS

Specifications

  • Flow rate:
    6,5 l/min
  • Description:
    Suction siphon pump
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

Suction operated siphon pump.

  • With PE suction tube 390 mm length
  • For drums up to 25 litres