ContactUsLinkComponent
Digital polarimeter, AP-300
Digital polarimeter, AP-300
Supplier:  Atago
Certificates
undefined
Digital polarimeter, AP-300
Supplier:  Atago
Specifications
  • Catalog No:
  • 635-0501
  • 635-0502
  • 635-0500
  • 635-0409
  • Wavelength:
  • 589 nm
  • Angular scale accuracy:
  • ±0,01 °A
  • Angular scale range:
  • −89,99…+89,9 °A
  • Angular scale resolution:
  • 0,01 °A
  • Sugar scale accuracy:
  • ±0,03 °Z
  • Sugar scale range:
  • −130…+130 °Z
  • Memory:
  • 30 values
  • Light source:
  • Halogen
  • Temperature compensation:
  • 18 - 30 °C
  • Temperature accuracy:
  • ±0,05 °C