Order Entry
Switzerland
ContactUsLinkComponent
Upright microscope, F1115
Upright microscope, F1115
Supplier:  MOTIC
Certificates
undefined
Upright microscope, F1115
Supplier:  MOTIC

Specifications

  • Catalog No:
  • 632-9061
  • 632-9062
  • 630-0701
  • 632-9059
  • Head:
  • 45° inclined, 360° rotating
  • Nosepiece:
  • Quadruple
  • Quadruple
  • Quadruple, Quadruple
  • Quadruple
  • Objectives:
  • Achromatic 4×, 10×, 40× (spring)
  • Stage:
  • 120×110 mm
  • Condenser:
  • Abbe 1,25 N.A. with iris diaphragm
  • Illumination:
  • LED
  • LED
  • LED
  • LED

Specifications

Related Information

Frequently Bought Together