Disposable tubes for ULTRA-TURRAX® Tube Drive and ULTRA-TURRAX® Tube Drive control
Supplier: IKA
Certificates
About this item
ST-20 and ST-50 tubes comprise of tube, stirring device and closure.
- Ideal for mixing, stirring and extractions
- Suitable for preparation of soil sample suspensions and solubility testing for pharmaceuticals
- 20 and 50 ml volume tubes
DT-20 and DT-50 tubes comprise of tube, dispersion device and closure.
Ideal for dispersion, homogenisation and suspensions
Suitable for use in pharmacokinetics, metabolism studies and diagnosis
20 and 50 ml volume tubes
BMT-20-G and BMT-50-G ball-mill tubes comprise of tube, ball (G = glass) and closure.
Ideal for dry milling of dry and brittle samples (e.g. kaolin, gypsum, coloured pigments, tablets)
Suitable for cell maceration and processing of materials mixed with fluids
20 and 50 ml volume tubes
Ordering information: Other tubes are available, including sterile versions (denoted as gamma) and tubes with pierceable membrane (M) closures for dosing and sampling.
Frequently Bought Together