Order Entry
Switzerland
ContactUsLinkComponent
Ultra-low temperature freezers, -86 °C, MDF chest models, PHCbi
Ultra-low temperature freezers, -86 °C, MDF chest models, PHCbi
Supplier:  PHC Corporation
Certificates
undefined
Ultra-low temperature freezers, -86 °C, MDF chest models, PHCbi
Supplier:  PHC Corporation

Specifications

  • Catalog No:
  • 472-0109
  • 471-0135
  • 471-0421
  • 471-0133
  • Model:
  • MDF-C8V (VIP Plus)
  • MDF-793
  • MDF-C8V1 (VIP)
  • MDF-393
  • Display:
  • LED
  • Digital
  • LED
  • Digital
  • Alarm:
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Door type:
  • 1 outer door, 1 inner door
  • 1 outer door, 4 inner doors
  • 1 outer door, 1 inner door
  • 1 outer door, 3 inner doors
  • Door lock:
  • Yes
  • Yes
  • Yes
  • Yes
  • Weight:
  • 67 kg
  • 334 kg
  • 67 kg
  • 247 kg

Specifications

Frequently Bought Together