ContactUsLinkComponent
CentriVap® mobile systems
CentriVap® mobile systems
Supplier:  LABCONCO
Certificates
CentriVap® mobile systems
Supplier:  LABCONCO
Specifications
  • Catalog No:
  • 531-0169
  • 531-0170
  • Alarm:
  • An audible alarm signals when the specified operating time has expired
  • An audible alarm signals when the specified operating time has expired
  • Heat range:
  • Off to HI (110 °C) in increments of 1°
  • Heating time:
  • 1-999 min
  • 1-999 min
  • Run time:
  • 1 - 999 min
  • 1 - 999 min