Electrodes, Genepaddles
Anbieter: BTX
Zertifikate
Über diesen Artikel
Genepaddles are non invasive, paddle-style, reusable electrodes designed for in vivo and in vitro applications, such as gene delivery (IVEGD) in mouse embryos. The electrodes are placed anterior and posterior to the embryo following injection of the molecule of interest, and an electroporation pulse is delivered.
- Gold plated rectangular paddles, available in two sizes 3×5 mm or 5×7 mm
- Genepaddles may be positioned in parallel at a predetermined gap in tissue using the Genetrode/Genepaddle holder
Generator compatibility: ECM® 830, ECM® 2001
Voltage range: 0 - 200 V DC (do not use AC)
Pulse length range: 10 µsec to 99 msec in PBS
Pulse number range: 1 - 99 (depending on voltage and pulse length settings)
Genetrode™ holder electrode gap: 1 - 10 mm range.
Requires Genetrodes/Genepaddle holder, square post cables and coaxial banana plug cable.
Genetrode™ holder electrode gap: 1 - 10 mm range
Frequently Bought Together