ContactUsLinkComponent
Oxygen meter, online, insert, Quadroline Oxi 296
Oxygen meter, online, insert, Quadroline Oxi 296
Catalog # 664-0015
Supplier:  WTW ONLINE
undefined
Oxygen meter, online, insert, Quadroline Oxi 296
Catalog # 664-0015
Supplier:  WTW ONLINE
Specifications
  • Description:
    Online oxygen meter, insert, Oxi 296
  • Pk:
    1
Specifications
About this item
Suitable for the most common applications in the water and waste water sectors.
  • With two relay contacts and an analogue output for oxygen and temperature readings
Ordering information: Electrodes with various cable lengths, frames and mounting kits for horizontal and vertical ducts available on request.