Orders LinkContactUsLinkComponent
Mechanical cell counter
Mechanical cell counter
Mechanical counter, 0 − 9999
Codice catalogo 609-1145
Nome del fornitore:   GLASWARENFABRIK KARL HECHT
Mechanical cell counter
Codice catalogo 609-1145
Nome del fornitore:  GLASWARENFABRIK KARL HECHT
Mechanical cell counter
Mechanical cell counter
Mechanical counter, 0 − 9999
Price
SFr. 35.20
Log in per disponibilità
1 * 1 ST
Product Details & Documents
This manual steel cell counter allows for a mechanical counting from 0 to 9999.
  • Finger lever
Colour: Chrome
Specifiche
  • Range:
    0 − 9999
  • Description:
    Mechanical counter