ContactUsLinkComponent
VWR®, Tubing, High Temperature for Circulators
VWR®, Tubing, High Temperature for Circulators
Catalog # 461-0106
Supplier:  VWR Collection
undefined
VWR®, Tubing, High Temperature for Circulators
Catalog # 461-0106
Supplier:  VWR Collection
Specifications
  • Description:
    High temperature tubing, for VWR and other circulators
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
PTFE lined, stainless steel braid.
Flexible PTFE lined, stainless steel over braided tubing with 6,3 mm NPT quick connectors at both ends. Maximum temperature is 300 °C.
Length (mm): 930