Rotors for Kr4i/Kr25i Centrifuges
Anbieter: JOUAN
Zertifikate
Über diesen Artikel
Utilize the rotating capabilities of the Thermo Scientific™ RP6 Windshielded Swinging-Bucket Rotor, a high-capacity, low-speed rotor.
Capacity: 6×1000 ml (6 L) with Round bucket, 12×250 ml (3 L) with Rectangular bucket, Max. speed/RCF: 4250 rpm/5,735×g with Round bucket, 5,392×g with Rectangular rotor.
Frequently Bought Together