Orders LinkContactUsLinkComponent
Counting chambers, Thoma
Counting chambers, Thoma
Counting chambers, Thoma
Counting chambers, Thoma
For counting thrombocytes and erythrocytes, the grid system corresponds to the large square in the middle of the Neubauer chamber.
  • Subdivision doubles the analysis options
  • Bright-line division increases contrast of the net system to make work much easier
  • Spring clips guarantee the exact positioning of the cover slips
  • For blood capillaries
  • Each small square has a surface of 0,0025 mm²
Product Family Options
  • Description
    Depth of chamber
  • Counting chamber with double ruling, Thoma
    0,10 mm
    1 * 1 ST
    1 * 1 ST - SFr. 140.00
    SFr. 140.00
    1 * 1 ST
       SFr. 140.00
  • Counting chamber with double ruling, bright-lined, Thoma
    0,10 mm
    1 * 1 ST
    1 * 1 ST - SFr. 128.00
    SFr. 128.00
    1 * 1 ST
       SFr. 128.00
  • Counting chamber with double ruling and clips, Thoma
    0,10 mm
    1 * 1 ST
    1 * 1 ST - SFr. 102.00
    SFr. 102.00
    1 * 1 ST
       SFr. 102.00
  • Cell counter with double ruling and clips, bright-lined, Thoma
    0,1 mm
    1 * 1 ST
    1 * 1 ST - SFr. 157.00
    SFr. 157.00
    1 * 1 ST
       SFr. 157.00