ContactUsLinkComponent
Portable autoclaves, MLS series, PHCbi
Portable autoclaves, MLS series, PHCbi
Supplier:  PHC Corporation
Certificates
undefined
Portable autoclaves, MLS series, PHCbi
Supplier:  PHC Corporation
Specifications
  • Catalog No:
  • 481-0687
  • 481-0507
  • Model:
  • MLS-3751L
  • MLS-3020U
  • Capacity:
  • 50 L
  • 48 L
  • Temperature:
  • 115...135 °C
  • 0-138 °C
  • Temperature range:
  • 0-138 °C
  • Int.Ø×H:
  • 370×415 mm
  • 300×670 mm
  • W×D×H ext.:
  • 478×632×748 mm
  • 440×550×1050 mm